missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10603793
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10603793 10 g Amber Glass Bottle 10g
10062214 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10603793 Supplier Thermo Scientific Maybridge Supplier No. EN00019EA
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 27006-76-4
Molecular Formula C6H7ClN2O
Molecular Weight (g/mol) 158.585
InChI Key SZRSMNYUEXXEBL-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole-4-carbaldehyde, 5-chloro-1,3-dimethyl-1h-pyrazole-4-carboxaldehyde, 5-chloro-1,3-dimethyl pyrazole-4-aldehyde, 5-chloro-1,3-dimethylpyrazole-4-carboxaldehyde, 5-chloro-4-formyl-1,3-dimethylpyrazole, 5-chloro-1,3-dimethyl-1h-pyrazole-4-aldehyde, 1h-pyrazole-4-carboxaldehyde, 5-chloro-1,3-dimethyl, pubchem23629, acmc-20a2nm, ksc496s9t
PubChem CID 2777397
IUPAC Name 5-chloro-1,3-dimethylpyrazole-4-carbaldehyde
SMILES CC1=NN(C(=C1C=O)Cl)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 158.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-1, 3-dimethyl-1H-pyrazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.