missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-1,3-dimethoxybenzene, 97%, Thermo Scientific™

Product Code. 10022500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10022500 5 g Glass Bottle 5g
10541164 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10022500 Supplier Thermo Scientific Acros Supplier No. 197770050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7051-16-3
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.608
MDL Number MFCD00008382
InChI Key WQHNWJBSROXROL-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethoxybenzene, 3,5-dimethoxychlorobenzene, benzene, 1-chloro-3,5-dimethoxy, 5-chlororesorcinol dimethyl ether, 1,3-dimethoxy-5-chlorobenzene, pubchem3042, benzene,5-dimethoxy, acmc-209oev, wln: gr co1 eo1, 3,5-dimethoxyphenyl chloride
PubChem CID 81502
IUPAC Name 1-chloro-3,5-dimethoxybenzene
SMILES COC1=CC(=CC(=C1)Cl)OC

Specifications

Melting Point 33.0°C to 37.0°C
CAS Min % 96.0
Color White to Yellow or Pink
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Crystalline Powder
Assay Percent Range 97%
Linear Formula ClC6H3(OCH3)2
Beilstein 06, III, 4335
Formula Weight 172.61
Percent Purity 97%
Chemical Name or Material 5-Chloro-1, 3-dimethoxybenzene
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chloro-1,3-dimethoxybenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.