missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,3-benzodioxole, 98%

Product Code. 11410747
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410747 50 g 50g
11400747 10 g 10g
11420747 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410747 Supplier Thermo Scientific Alfa Aesar Supplier No. B22534.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7228-38-8 | C7H5ClO2 | 156.565 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7228-38-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00010842
InChI Key ODQPZHOXLYATLC-UHFFFAOYSA-N
PubChem CID 138966
IUPAC Name 5-chloro-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)Cl

Specifications

Density 1.34
Boiling Point 185°C to 187°C
Flash Point 93°C (201°F)
Refractive Index 1.556
Quantity 50 g
Synonym 5-chlorobenzo d 1,3 dioxole, 1-chloro-3,4-methylenedioxybenzene, 1,3-benzodioxole, 5-chloro, 5-chloro-2h-1,3-benzodioxole, 1,3-benzodioxole,5-chloro, 5-chloro-benzo 1,3 dioxole, pubchem3620, 4-chloro-1,2-methylenedioxybenzene, 5-chloro-1,3-benzodioxole, ukrorgsyn-bb bbr-008667
Formula Weight 156.57
Percent Purity 98%
Chemical Name or Material 5-Chloro-1,3-benzodioxole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.