missing translation for 'onlineSavingsMsg'
Learn More

(5-Carboxypentyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Product Code. 11408997
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408997 1 g 1g
11418997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408997 Supplier Thermo Scientific Alfa Aesar Supplier No. B20588.03
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

It is used in medicine and as pharmaceutical intermediate. It is also employed as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine and as pharmaceutical intermediate. It is also employed as catalyst.

Solubility
Soluble in Chloroform and Methanol.

Notes
Hygroscopic. Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50889-29-7
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00055556
InChI Key JUWYRPZTZSWLCY-UHFFFAOYSA-N
Synonym 5-carboxypentyl triphenylphosphonium bromide, 5-carboxypentyl triphenyl phosphonium bromide, 5-carboxypentyl triphenylphosphanium bromide, 5-carboxypentyltriphenylphosphonium bromide, phosphonium, 5-carboxypentyl triphenyl-, bromide, acmc-20apmi, pxjvnuuwrjumaq-uhfffaoysa-m, 5-carboxyamyl triphenylphosphonium bromide
PubChem CID 2779280
IUPAC Name 5-carboxypentyl(triphenyl)phosphanium;bromide
SMILES [Br-].OC(=O)CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 200°C to 202°C
Quantity 1 g
Solubility Information Soluble in Chloroform and Methanol.
Formula Weight 457.36
Percent Purity 97%
Chemical Name or Material (5-Carboxypentyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.