missing translation for 'onlineSavingsMsg'
Learn More

5-Carboxyfluorescein Dipivalate N-Hydroxysuccinimide Ester, TRC

Product Code. 30357736
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357736 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357736 Supplier Toronto Research Chemicals Supplier No. TRCC1783531MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Carboxyfluorescein Dipivalate N-Hydroxysuccinimide Ester
CAS 186032-65-5
Molecular Formula C35 H31 N O11
Quantity 1 mg
Synonym 5-Carboxyfluorescein Dipivalate NHS Ester, 5-Carboxyfluorescein Dipivalate Succinimide Ester, 3',6'-Bis(2,2-dimethyl-1-oxopropoxy)-3-oxo-spiro[isobenzofuran-1(3H),9'-[9H]xanthene]-5-carboxylic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, 5-(((2,5-Dioxopyrrolidin-1-yl)oxy)carbonyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl Bis(2,2-dimethylpropanoate)
InChI Formula InChI=1S/C35H31NO11/c1-33(2,3)31(41)43-19-8-11-23-25(16-19)45-26-17-20(44-32(42)34(4,5)6)9-12-24(26)35(23)22-10-7-18(15-21(22)30(40)46-35)29(39)47-36-27(37)13-14-28(36)38/h7-12,15-17H,13-14H2,1-6H3
SMILES CC(C)(C)C(=O)Oc1ccc2c(Oc3cc(OC(=O)C(C)(C)C)ccc3C24OC(=O)c5cc(ccc45)C(=O)ON6C(=O)CCC6=O)c1
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3',6'-bis(2,2-dimethylpropanoyloxy)-3-oxo-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate
Molecular Weight (g/mol) 641.62
Formula Weight 641.1897
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.