missing translation for 'onlineSavingsMsg'
Learn More

5-Carbethoxyuracil, 98%, Thermo Scientific™

Product Code. 10581774 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10581774 25 g 25g
10042842 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10581774 Supplier Thermo Scientific Acros Supplier No. 373970250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 28485-17-8
Molecular Formula C7H8N2O4
Molecular Weight (g/mol) 184.15
MDL Number MFCD00057337
InChI Key MKNYHTGOVKPZMU-UHFFFAOYSA-N
Synonym 5-carbethoxyuracil, ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, isoorotic acid ethyl ester, ethyl 2,4-dihydroxypyrimidine-5-carboxylate, ethyl uracil-5-carboxylate, ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester, ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate, pubchem13254
PubChem CID 101411
IUPAC Name ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CNC(=O)NC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 232.0°C to 235.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 184.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Carbethoxyuracil
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.