missing translation for 'onlineSavingsMsg'
Learn More

5-Carbamoylpyridine-3-boronic acid pinacol ester, 96%

Product Code. p-7034640
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429745 250 mg 250mg
15419745 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429745 Supplier Thermo Scientific Alfa Aesar Supplier No. H52330.MD
Add to basket
This item is not returnable. View return policy

CAS: 1169402-51-0 | C12H17BN2O3 | 248.089 g/mol

5-Carbamoylpyridine-3-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Carbamoylpyridine-3-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1169402-51-0
Molecular Formula C12H17BN2O3
Molecular Weight (g/mol) 248.089
MDL Number MFCD11878274
InChI Key NJLZODHYCOZPBJ-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinamide, 5-carbamoylpyridine-3-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carboxamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-nicotinamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl nicotinamide, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carboxamide, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinecarboxamide
PubChem CID 59023117
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)C(=O)N

Specifications

Quantity 250 mg
Formula Weight 248.09
Percent Purity 96%
Chemical Name or Material 5-Carbamoylpyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.