missing translation for 'onlineSavingsMsg'
Learn More

5-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15484916
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484916 1 g 1g
15494916 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484916 Supplier Thermo Scientific Alfa Aesar Supplier No. H59262.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1412905-42-0
Molecular Formula C13H17BFNO3
Molecular Weight (g/mol) 265.09
MDL Number MFCD18730404
InChI Key RBEAFMIZDXOHAT-UHFFFAOYSA-N
Synonym 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 5-carbamoyl-2-fluorobenzeneboronic acid pinacol ester
PubChem CID 99738340
IUPAC Name 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O

Specifications

Quantity 1 g
Formula Weight 265.09
Percent Purity 96%
Chemical Name or Material 5-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.