missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress 5-BrUTP sodium salt
Specifications
Specifications
| Product Type | Biochemical Assay Reagents |
| Content And Storage | -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| Shipping Condition | Room temperature |
| Purity | 94.0% |
| Quantity | 5 mg |
| Solubility Information | H2O : 33.33 mg/mL (Need ultrasonic) |
| CAS | 161848-60-8 |
| Synonym | 5-Bromouridine 5'-triphosphate sodium salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?