missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromovanillin, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 5-Bromovanillin |
| CAS | 2973-76-4 |
| Molecular Formula | C8 H7 Br O3 |
| Quantity | 5 g |
| Synonym | 3-Bromo-4-hydroxy-5-methoxybenzaldehyde, 3-Bromo-5-methoxy-4-hydroxybenzaldehyde, 3-Methoxy-4-hydroxy-5-bromobenzaldehyde, 5-Bromo-3-methoxy-4-hydroxybenzaldhyde, 5-Bromo-4-hydroxy-3-anisaldehyde, 5-Bromo-4-hydroxy-3-methoxybenzaldehyde, 5-Bromovanillin, 6-Bromo-4-formyl-2-methoxyphenol, NSC 10073, NSC 29037 |
| InChI Formula | InChI=1S/C8H7BrO3/c1-12-7-3-5(4-10)2-6(9)8(7)11/h2-4,11H,1H3 |
| SMILES | COc1cc(C=O)cc(Br)c1O |
| IUPAC Name | 3-bromo-4-hydroxy-5-methoxy-benzaldehyde |
| Molecular Weight (g/mol) | 231.04 |
| Formula Weight | 229.9579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction