missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovanillin, 98%

Product Code. 11435283
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11435283

Brand: Thermo Scientific Alfa Aesar A12861.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2973-76-4 | C8H7BrO3 | 231.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2973-76-4
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD00006940
InChI Key KLSHZDPXXKAHIJ-UHFFFAOYSA-N
Synonym 5-bromovanillin, benzaldehyde, 3-bromo-4-hydroxy-5-methoxy, vanillin, 5-bromo, 5-bromo-4-hydroxy-3-methoxybenzaldehyde, 3-bromo-4-hydroxy-5-methoxy-benzaldehyde, unii-508wvs56pw, 3-bromo-4-hydroxy-5-methyloxy benzaldehyde, 5-bromovaniline, 5-bromovanilline
PubChem CID 18099
IUPAC Name 3-bromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(Br)=C1O

Specifications

Melting Point 162°C to 166°C
Quantity 100 g
Beilstein 2049761
Sensitivity Air and light sensitive
Formula Weight 231.05
Percent Purity 98%
Chemical Name or Material 5-Bromovanillin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.