missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromovaleric acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
250 g
Description
5-Bromovaleric acid is used in the synthesis of inhibitors for aminoglycoside resistant bacteria. 5-Bromovaleric acid is also used in the preparation of sequence specific base pair mimics as topoisomerase IB inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromovaleric acid is used in the synthesis of inhibitors for aminoglycoside resistant bacteria. 5-Bromovaleric acid is also used in the preparation of sequence specific base pair mimics as topoisomerase IB inhibitors.
Solubility
Soluble in ethanol. Insoluble in water.
Notes
Store away from strong bases and oxidizing agents. Store at room temperature.
5-Bromovaleric acid is used in the synthesis of inhibitors for aminoglycoside resistant bacteria. 5-Bromovaleric acid is also used in the preparation of sequence specific base pair mimics as topoisomerase IB inhibitors.
Solubility
Soluble in ethanol. Insoluble in water.
Notes
Store away from strong bases and oxidizing agents. Store at room temperature.
Specifications
Melting Point | 38°C to 41°C |
Boiling Point | 142°C to 145°C (13 mmHg) |
Flash Point | >100°C (212°F) |
Quantity | 250 g |
UN Number | UN3261 |
Beilstein | 1745664 |
Solubility Information | Soluble in ethanol. Insoluble in water. |
Formula Weight | 181.03 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromovaleric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'