missing translation for 'onlineSavingsMsg'
Learn More

5-Bromouridine, 98%

Product Code. 11315199
Click to view available options
Quantity:
0.25 g
Unit Size:
0.25g
This item is not returnable. View return policy

Product Code. 11315199

Brand: Thermo Scientific Alfa Aesar A18507.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 957-75-5 | C9H11BrN2O6 | 323.10 g/mol

5-Bromouridine is often incorporated into RNA in order to detect immunocytochemically and analyzed by cytometry. It possesses anti-viral activity and inhibits the activity of viruses such as human immunodeficiency virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromouridine is often incorporated into RNA in order to detect immunocytochemically and analyzed by cytometry. It possesses anti-viral activity and inhibits the activity of viruses such as human immunodeficiency virus.

S
TRUSTED_SUSTAINABILITY

Specifications

CAS 957-75-5
Melting Point 191°C to 193°C
Molecular Formula C9H11BrN2O6
MDL Number MFCD00006528
Quantity 0.25 g
Synonym 5-bromouridine, uridine, 5-bromo, 1-beta-ribofuranosyl-5-bromo-uracil, 5-bromo-1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, bromouridine, 5-bromo-uridine, key8pg1brc, unii-key8pg1brc, 5-bromouracil ribonucleoside, bru
Solubility Information Insoluble in water.
InChI Key AGFIRQJZCNVMCW-UHFFFAOYNA-N
SMILES OCC1OC(C(O)C1O)N1C=C(Br)C(=O)NC1=O
IUPAC Name 5-bromo-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 323.10
PubChem CID 91494
ChEBI CHEBI:20553
Formula Weight 323.1
Percent Purity 98%
Chemical Name or Material 5-Bromouridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.