missing translation for 'onlineSavingsMsg'
Learn More

5-Bromouracil, 98+%

Product Code. 11482537
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482537 100 g 100g
11462537 5 g 5g
11472537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482537 Supplier Thermo Scientific Alfa Aesar Supplier No. A14799.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-20-7 | C4H3BrN2O2 | 190.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-20-7
Molecular Formula C4H3BrN2O2
Molecular Weight (g/mol) 190.98
MDL Number MFCD00006017
InChI Key LQLQRFGHAALLLE-UHFFFAOYSA-N
Synonym 5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 5802
ChEBI CHEBI:20552
IUPAC Name 5-bromo-1H-pyrimidine-2,4-dione
SMILES BrC1=CNC(=O)NC1=O

Specifications

Melting Point ∼310°C (decomposition)
Quantity 100 g
Beilstein 127176
Merck Index 14,1440
Formula Weight 190.99
Percent Purity ≥98%
Chemical Name or Material 5-Bromouracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.