missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Bromouracil, 98%

Product Code. 10647212 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Packaging:
Glass bottle
missing translation for 'unitSize'
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10647212 5 g Glass bottle 5g
11370636 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10647212 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 226440050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-20-7 | C4H3BrN2O2 | 190.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-20-7
Molecular Formula C4H3BrN2O2
Molecular Weight (g/mol) 190.98
MDL Number MFCD00006017
InChI Key LQLQRFGHAALLLE-UHFFFAOYSA-N
Synonym 5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 5802
ChEBI CHEBI:20552
IUPAC Name 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES BrC1=CNC(=O)NC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 307.0°C to 310.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 24,318
Merck Index 15,145
Solubility Information Solubility in water: soluble in cold water. Other solubilities: soluble in ammonia water and other alkalines
Formula Weight 190.98
Percent Purity 98%
Chemical Name or Material 5-Bromouracil, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Bromouracil
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.