missing translation for 'onlineSavingsMsg'
Learn More

5-Bromosalicylaldehyde, 99%

Product Code. 10592171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10592171 100 g Glass bottle 100g
10113070 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10592171 Supplier Thermo Scientific Acros Supplier No. 159911000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1761-61-1 | C7H5BrO2 | 201.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1761-61-1
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00003330
InChI Key MKKSTJKBKNCMRV-UHFFFAOYSA-N
Synonym 5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779
PubChem CID 72863
IUPAC Name 5-bromo-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 103°C to 107°C
Color Beige-Yellow to Yellow
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H3(OH)CHO
Quantity 100 g
Beilstein 08, 54
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 201.02
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Bromosalicylaldehyde, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.