missing translation for 'onlineSavingsMsg'
Learn More

5-Bromosalicylaldehyde, 98%

Product Code. 11423904
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423904 100 g 100g
11413904 25 g 25g
11433904 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423904 Supplier Thermo Scientific Alfa Aesar Supplier No. A11917.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1761-61-1 | C7H5BrO2 | 201.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1761-61-1
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00003330
InChI Key MKKSTJKBKNCMRV-UHFFFAOYSA-N
Synonym 5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779
PubChem CID 72863
IUPAC Name 5-bromo-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)O

Specifications

Melting Point 103°C to 106°C
Quantity 100 g
Beilstein 774710
Sensitivity Air Sensitive
Formula Weight 201.02
Percent Purity 98%
Chemical Name or Material 5-Bromosalicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.