missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoquinoline, 97%, Thermo Scientific Chemicals

Product Code. 11355425
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11355425 Supplier Thermo Scientific Alfa Aesar Supplier No. L19634.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4964-71-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00234481
InChI Key CHODTZCXWXCALP-UHFFFAOYSA-N
Synonym 5-bromo-quinoline, quinoline, 5-bromo, 5-bromooquinoline, 5-bromo quinoline, 5-bromoquinolline, pubchem7116, 5-bromoquinoline, acmc-209kh7, ksc492i0h, 5-bromoquinoline 250mg
PubChem CID 817321
IUPAC Name 5-bromoquinoline
SMILES C1=CC2=C(C=CC=N2)C(=C1)Br

Specifications

Melting Point 44°C to 48°C
Boiling Point 105°C to 107°C (1 mmHg)
Quantity 1 g
Beilstein 115148
Formula Weight 208.06
Percent Purity 97%
Chemical Name or Material 5-Bromoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.