missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyrimidine, 98%

Product Code. 10511651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10511651 25 g Glass bottle 25g
10521651 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10511651 Supplier Thermo Scientific Acros Supplier No. 204300250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4595-59-9 | C4H3BrN2 | 158.986 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4595-59-9
MDL Number MFCD00006117
InChI Key GYCPLYCTMDTEPU-UHFFFAOYSA-N
Synonym pyrimidine, 5-bromo, 5-bromo pyrimidine, 5-bromo-1,3-diazine, 5-bromo-pyrimidine, 5-brpy compound, 5-brompyrimidin, 5-brornopyrimidine, zlchem 349, pubchem6902, 11-cis-retinyl-palmitate
PubChem CID 78344
IUPAC Name 5-bromopyrimidine
SMILES C1=C(C=NC=N1)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H3BrN2
Linear Formula (C5H3N2)Br
Quantity 25 g
Molecular Weight (g/mol) 158.99
Formula Weight 158.99
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 5-Bromopyrimidine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromopyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.