Learn More
5-Bromopyridine-3-carbohydrazide, 97%, Maybridge™
Brand: Maybridge RDP00027EA
Code : 31
Additional Details : CAS Number : 112193-41-6 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 10g |
Chemical Identifiers
112193-41-6 | |
216.038 | |
VERXTAUONOFMLH-UHFFFAOYSA-N | |
696365 | |
C1=C(C=NC=C1Br)C(=O)NN |
C6H6BrN3O | |
MFCD00173916 | |
5-bromonicotinohydrazide, 3-pyridinecarboxylicacid, 5-bromo-, hydrazide, bromonicotinohydrazide, acmc-20ea1n, maybridge1_006123, 5-bromonicotinic acid hydrazide, 5-bromo-3-pyridinecarbohydrazide, 5-bromo-nicotinic acid hydrazide, 3-pyridinecarboxylicacid,5-bromo-,hydrazide, 3-pyridinecarboxylic acid, 5-bromo-, hydrazide | |
5-bromopyridine-3-carbohydrazide |
Specifications
5-Bromopyridine-3-carbohydrazide | |
10g | |
97.0 | |
C6H6BrN3O | |
5-bromonicotinohydrazide, 3-pyridinecarboxylicacid, 5-bromo-, hydrazide, bromonicotinohydrazide, acmc-20ea1n, maybridge1_006123, 5-bromonicotinic acid hydrazide, 5-bromo-3-pyridinecarbohydrazide, 5-bromo-nicotinic acid hydrazide, 3-pyridinecarboxylicacid,5-bromo-,hydrazide, 3-pyridinecarboxylic acid, 5-bromo-, hydrazide | |
C1=C(C=NC=C1Br)C(=O)NN | |
216.038 | |
216.04 |
Amber Glass Bottle | |
112193-41-6 | |
97% | |
MFCD00173916 | |
VERXTAUONOFMLH-UHFFFAOYSA-N | |
5-bromopyridine-3-carbohydrazide | |
696365 | |
97% |