missing translation for 'onlineSavingsMsg'
Learn More

(5-Bromopyrid-2-yl)methanol, 97%, Thermo Scientific™

Product Code. 10235414
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10235414 1 g 1g
10577854 250 mg 250mg
10113084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10235414 Supplier Thermo Scientific Maybridge Supplier No. MO07897DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 88139-91-7
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD04035597
InChI Key RUCZFWMEACWFER-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxymethylpyridine, 2-hydroxymethyl-5-bromopyridine, 5-bromopyridin-2-yl methanol, 5-bromo-2-hydroxymethyl pyridine, 5-bromopyrid-2-yl methanol, 5-bromo-2-pyridinemethanol, 5-bromo-pyridin-2-yl-methanol, 2-pyridinemethanol, 5-bromo, 5-bromo-2-pyridyl methan-1-ol, 5-bromopyridine-2-methanol
PubChem CID 5200169
IUPAC Name (5-bromopyridin-2-yl)methanol
SMILES OCC1=CC=C(Br)C=N1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 188.02
Percent Purity 97%
Chemical Name or Material (5-Bromopyrid-2-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.