missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopentyl Acetate, TRC

Product Code. 30338214
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338214 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338214 Supplier Toronto Research Chemicals Supplier No. TRCB68610050G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Bromopentyl Acetate
CAS 15848-22-3
Molecular Formula C7H13BrO2
Quantity 50 g
Synonym 5-Bromo-1-pentanol Acetate, 1-Acetoxy-5-bromopentane, 5-Bromo-1-acetoxypentane
InChI Formula InChI=1S/C7H13BrO2/c1-7(9)10-6-4-2-3-5-8/h2-6H2,1H3
SMILES CC(=O)OCCCCCBr
IUPAC Name 5-bromopentyl acetate
Molecular Weight (g/mol) 209.08
Formula Weight 208.0099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.