missing translation for 'onlineSavingsMsg'
Learn More

5-Bromooxindole, 98%

Product Code. 11420050
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420050 5 g 5g
11410050 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420050 Supplier Thermo Scientific Alfa Aesar Supplier No. H50573.06
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 20870-78-4 | C8H6BrNO | 212.046 g/mol

5-Bromooxindole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromooxindole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in Dimethylformamide.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20870-78-4
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD00456998
InChI Key VIMNAEVMZXIKFL-UHFFFAOYSA-N
Synonym 5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
PubChem CID 611193
IUPAC Name 5-bromo-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)Br)NC1=O

Specifications

Quantity 5 g
Solubility Information Soluble in Dimethylformamide.
Formula Weight 212.05
Percent Purity 98%
Chemical Name or Material 5-Bromooxindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.