missing translation for 'onlineSavingsMsg'
Learn More

5-Bromonicotinonitrile, 97%, Thermo Scientific™

Product Code. 10123774
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10123774 25 g 25g
10551492 1 g 1g
10602453 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10123774 Supplier Thermo Scientific Maybridge Supplier No. KM08398EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 35590-37-5
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
MDL Number MFCD00174363
InChI Key FTFFHWWIPOQCBC-UHFFFAOYSA-N
Synonym 5-bromonicotinonitrile, 3-bromo-5-cyanopyridine, 5-bromo-3-cyanopyridine, 3-cyano-5-bromopyridine, 3-pyridinecarbonitrile, 5-bromo, 3-bromo-5-cyano-pyridine, 5-bromo-3-pyridinecarbonitrile, pubchem7702, acmc-209ihz, ksc492g7p
PubChem CID 736793
IUPAC Name 5-bromopyridine-3-carbonitrile
SMILES BrC1=CN=CC(=C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 183.01
Percent Purity 97%
Chemical Name or Material 5-Bromonicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.