missing translation for 'onlineSavingsMsg'
Learn More

5-Bromonicotinic acid, 98%

Product Code. 10246650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246650 10 g Glass bottle 10g
10358490 50 g Glass bottle 50g
10647304 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10246650 Supplier Thermo Scientific Acros Supplier No. 211390100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20826-04-4 | C6H4BrNO2 | 202.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20826-04-4
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00009783
InChI Key FQIUCPGDKPXSLL-UHFFFAOYSA-N
Synonym 5-bromonicotinic acid, 5-bromo nicotinic acid, 5-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo, nicotinic acid, 5-bromo, 5-bromo-nicotinic acid, 3-bromo-5-pyridine carboxylic acid, 3-bromo pyridine-5-carboxylic acid, pubchem1211, acmc-209fev
PubChem CID 88707
IUPAC Name 5-bromopyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=CC(Br)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 178°C to 180°C
Color Brown or Gray-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula Br(C5H3N)COOH
Quantity 10 g
Beilstein 22, 44
Formula Weight 202.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.