missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Bromomethyl)-2,4-diphenyl-1,3-thiazole, Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 80.0 |
CAS Max % | 100.0 |
Quantity | 1 g |
Formula Weight | 330.25 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 5-(Bromomethyl)-2, 4-diphenyl-1, 3-thiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'