missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromomethyl)-2,1,3-benzoxadiazole, 97%, Thermo Scientific™

Product Code. 10562832
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 32863-31-3
Molecular Formula C7H5BrN2O
Molecular Weight (g/mol) 213.03
MDL Number MFCD00174369
InChI Key BUJFLTNYWUEROF-UHFFFAOYSA-N
Synonym 5-bromomethyl-2,1,3-benzoxadiazole, 5-bromomethyl benzo c 1,2,5 oxadiazole, 5-bromomethylbenzofurazan, 5-bromomethyl benzofurazan, 2,1,3-benzoxadiazole, 5-bromomethyl, zlchem 1034, 5-bromomethylbenzofurazane, 5-bromomethyl benzo c furazan, 5-bromomethyl-benzo 1,2,5 oxadiazole, 5-bromomethyl-2,1,3-benzooxadiazole
PubChem CID 2735587
IUPAC Name 5-(bromomethyl)-2,1,3-benzoxadiazole
SMILES BrCC1=CC2=NON=C2C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 213.03
Percent Purity 97%
Chemical Name or Material 5-(Bromomethyl)-2, 1, 3-benzoxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'