missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Bromomethyl)-2,1,3-benzothiadiazole, 95+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
Synonym | 5-bromomethyl-2,1,3-benzothiadiazole, 5-bromomethyl benzo c 1,2,5 thiadiazole, 5-bromomethylbenzo-2,1,3-thiadiazole, 5-bromomethyl-benzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole,5-bromomethyl, 2,1,3-benzothiadiazole, 5-bromomethyl, 5-bromomethyl benzo c 1,2,5-thiadiazole, 5-bromomethyl-2,1,3-benzthiadiazole, 5-bromomethylbenzo-2.1.3-thiadiazole, 5-bromomethylbenzo 2,1,3 thiadiazole |
Formula Weight | 229.1 |
Percent Purity | 95% |
Chemical Name or Material | 5-(Bromomethyl)-2, 1, 3-benzothiadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'