missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromoisoquinoline, 95%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Specifications
CAS | 34784-04-8 |
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Molecular Formula | C9H6BrN |
MDL Number | MFCD01646405 |
Quantity | 1g |
Synonym | isoquinoline, 5-bromo, 5-bromo-isoquinoline, 5-bromo isoquinoline, 5-bromoisoquinoline?, pubchem6214, acmc-1adiy, 5-bromoisoquinoline, ksc226k2l, buttpark 44\07-76, 5-bromoisoquinoline 250mg |
InChI Key | CYJZJGYYTFQQBY-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'