missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoisatin, 90+%

Product Code. 11450224
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450224 500 g 500g
11334669 50 g 50g
11440224 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450224 Supplier Thermo Scientific Alfa Aesar Supplier No. A13641.36
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87-48-9 | C8H4BrNO2 | 226.029 g/mol

5-Bromoisatin is used in the synthesis of N-derivatives of 5-bromoisatin, N-substituted pyrroles, linear polyaryleneoxindoles, 5-bromodioxindole, cinchoninic acid derivatives, 3-hydroxyoxindole, S-benzyldithiocarbazate Schiff Bases, 5-bromooxindole and Morita-Baylis-Hillman adducts of isatin derivatives. It is an indole derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromoisatin is used in the synthesis of N-derivatives of 5-bromoisatin, N-substituted pyrroles, linear polyaryleneoxindoles, 5-bromodioxindole, cinchoninic acid derivatives, 3-hydroxyoxindole, S-benzyldithiocarbazate Schiff Bases, 5-bromooxindole and Morita-Baylis-Hillman adducts of isatin derivatives. It is an indole derivative.

Solubility
Soluble in N,N dimethylformamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-48-9
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00149345
InChI Key MBVCESWADCIXJN-UHFFFAOYSA-N
Synonym 5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate
PubChem CID 6889
IUPAC Name 5-bromo-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1Br)C(=O)C(=O)N2

Specifications

Melting Point ∼250°C (decomposition)
Quantity 500 g
Beilstein 383760
Solubility Information Soluble in N,N dimethylformamide.
Formula Weight 226.03
Percent Purity ≥90%
Chemical Name or Material 5-Bromoisatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.