missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindoxyl diacetate, 98+%, Thermo Scientific Chemicals

Product Code. 11337626
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337626 1 g 1g
11347626 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337626 Supplier Thermo Scientific Alfa Aesar Supplier No. L11189.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33588-54-4 | Molecular Formula: C12H10BrNO3 | Molecular Weight: 296.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33588-54-4
Molecular Formula C12H10BrNO3
Molecular Weight (g/mol) 296.12
MDL Number MFCD00005799
InChI Key XJRIDJAGAYGJCK-UHFFFAOYSA-N
Synonym 5-bromoindoxyl diacetate, 1-acetyl-5-bromo-1h-indol-3-yl acetate, 5-bromoindoxyldiacetate, n-acetyl-5-bromoindolyl acetate, 1-acetyl-3-acetyloxy-5-bromoindole, 3-acetoxy-1-acetyl-5-bromoindole, 1h-indol-3-ol, 1-acetyl-5-bromo-, acetate ester, 3-acetoxy-1-acetyl-5-bromo-indole, 1h-indol-3-ol, 1-acetyl-5-bromo-, acetate, pubchem16263
PubChem CID 118505
IUPAC Name (1-acetyl-5-bromoindol-3-yl) acetate
SMILES CC(=O)N1C=C(C2=C1C=CC(=C2)Br)OC(=O)C

Specifications

Melting Point 122°C to 125°C
Quantity 1 g
Beilstein 236935
Formula Weight 296.13
Percent Purity ≥98%
Chemical Name or Material 5-Bromoindoxyl diacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.