missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindoline, TRC

Product Code. 30338754
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338754 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338754 Supplier Toronto Research Chemicals Supplier No. TRCB6846102G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Bromoindoline
CAS 22190-33-6
Molecular Formula C8H8BrN
Quantity 2 g
Synonym 5-Bromo-2,3-dihydro-1H-indole, 5-Bromo-2,3-dihydroindole
InChI Formula InChI=1S/C8H8BrN/c9-7-1-2-8-6(5-7)3-4-10-8/h1-2,5,10H,3-4H2
SMILES Brc1ccc2NCCc2c1
IUPAC Name 5-bromo-2,3-dihydro-1H-indole
Molecular Weight (g/mol) 198.06
Formula Weight 196.984
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.