missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindoline, 98+%

Product Code. 11383886
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383886 5 g 5g
11373886 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383886 Supplier Thermo Scientific Alfa Aesar Supplier No. L16992.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22190-33-6 | C8H8BrN | 198.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22190-33-6
Molecular Formula C8H8BrN
Molecular Weight (g/mol) 198.063
MDL Number MFCD00027410
InChI Key QEDCHCLHHGGYBT-UHFFFAOYSA-N
Synonym 5-bromoindoline, 1h-indole, 5-bromo-2,3-dihydro, 5-bromo-indoline, zlchem 28, 5-bromodihydroindole, pubchem7319, acmc-1cez6, 5-bromo-2,3-dihydro-indole, ksc497i6r
PubChem CID 3411566
IUPAC Name 5-bromo-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)Br

Specifications

Melting Point 36°C to 40°C
Boiling Point 114°C to 115°C (5 mmHg)
Quantity 5 g
Beilstein 121590
Formula Weight 198.06
Percent Purity ≥98%
Chemical Name or Material 5-Bromoindoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.