missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindoline, 97%, Thermo Scientific™

Product Code. 10604935 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10604935 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10604935 Supplier Thermo Scientific Acros Supplier No. 398630010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 22190-33-6
Boiling Point 114.0°C to 115.0°C (5.0 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8BrN
MDL Number MFCD00027410
Quantity 1g
Synonym 5-bromoindoline, 1h-indole, 5-bromo-2,3-dihydro, 5-bromo-indoline, zlchem 28, 5-bromodihydroindole, pubchem7319, acmc-1cez6, 5-bromo-2,3-dihydro-indole, ksc497i6r
Solubility Information Solubility in water: insoluble
InChI Key QEDCHCLHHGGYBT-UHFFFAOYSA-N
SMILES C1CNC2=C1C=C(C=C2)Br
IUPAC Name 5-bromo-2,3-dihydro-1H-indole
Molecular Weight (g/mol) 198.06
PubChem CID 3411566
Formula Weight 198.06
Percent Purity 97%
Chemical Name or Material 5-Bromoindoline
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromoindoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.