missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-3-carboxaldehyde, 97%

Product Code. 10706485 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10706485 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10706485 Supplier Thermo Scientific Acros Supplier No. 399790050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 877-03-2 | C9H6BrNO | 224.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 877-03-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 205°C to 206°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H6BrNO
MDL Number MFCD00152016
Quantity 5 g
Synonym 5-bromoindole-3-carboxaldehyde, 5-bromoindole-3-carbaldehyde, 5-bromoindole-3-carboxyaldehyde, 5-bromo-3-formylindole, 5-bromoindole-3-aldehyde, 5-bromo-3-formyl-1h-indole, 5-bromo-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 5-bromo, pubchem9122, acmc-209qpj
InChI Key PEENKJZANBYXNB-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1Br)C(=CN2)C=O
IUPAC Name 5-bromo-1H-indole-3-carbaldehyde
Molecular Weight (g/mol) 224.05
PubChem CID 70137
Formula Weight 224.05
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole-3-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromoindole-3-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.