missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromofuran-2-carboxaldehyde, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 5-Bromofuran-2-carboxaldehyde |
| CAS | 1899-24-7 |
| Molecular Formula | C5H3BrO2 |
| Quantity | 1 g |
| Synonym | 5-Bromo--2-furaldehyde, 2-Bromo-5-furancarboxaldehyde, 2-Bromofuran-5-carboxaldehyde, 5-Bromo-2-formylfuran, 5-Bromo-2-furaldehyde, 5-Bromo-2-furancarboxaldehyde, 5-Bromo-2-furfural, 5-Bromo-2-furylaldehyde, 5-Bromofuran-2-aldehyde, 5-Bromofurfural |
| InChI Formula | InChI=1S/C5H3BrO2/c6-5-2-1-4(3-7)8-5/h1-3H |
| SMILES | Brc1oc(C=O)cc1 |
| IUPAC Name | 5-bromofuran-2-carbaldehyde |
| Molecular Weight (g/mol) | 174.98 |
| Formula Weight | 173.9316 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction