missing translation for 'onlineSavingsMsg'
Learn More

5-Bromocytosine, 99%, Thermo Scientific Chemicals

Product Code. 10164320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164320 250 mg Glass bottle 250mg
10041900 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10164320 Supplier Thermo Scientific Acros Supplier No. 165782500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2240-25-7
Molecular Formula C4H4BrN3O
Molecular Weight (g/mol) 190.00
MDL Number MFCD00056025
InChI Key QFVKLKDEXOWFSL-UHFFFAOYSA-N
Synonym 5-bromocytosine, 4-amino-5-bromopyrimidin-2 1h-one, 6-amino-5-bromopyrimidin-2 1h-one, 4-amino-5-bromo-2-pyrimidinol, 4-amino-5-bromopyrimidin-2-ol, 2 1h-pyrimidinone, 4-amino-5-bromo, 4-amino-5-bromo-2-hydroxypyrimidine, 2 1h-pyrimidinone,6-amino-5-bromo, 6-amino-5-bromo-1,2-dihydropyrimidin-2-one, 4-amino-5-bromo-1,2-dihydropyrimidin-2-one
PubChem CID 75233
IUPAC Name 6-amino-5-bromo-1H-pyrimidin-2-one
SMILES NC1=C(Br)C=NC(=O)N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 240.0°C to 243.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 250 mg
Beilstein 24, 319
Formula Weight 190
Percent Purity 99%
Physical Form Amorphous Powder
Chemical Name or Material 5-Bromocytosine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Bromocytosine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.