missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 240.0°C to 243.0°C |
| Color | Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Beilstein | 24, 319 |
| Formula Weight | 190 |
| Show More |
Safety and Handling
Product Identifier
- 5-Bromocytosine
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?