missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-m-xylene, 98%

Product Code. 11974521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11974521 100 g Glass bottle 100g
10636671 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11974521 Supplier Thermo Scientific Acros Supplier No. 294001000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 556-96-7 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-96-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
InChI Key LMFRTSBQRLSJHC-UHFFFAOYSA-N
Synonym 5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene
PubChem CID 136357
IUPAC Name 1-bromo-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)Br)C

Specifications

Color Colorless to Yellow
Density 1.3600g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5475 to 1.5495
Assay Percent Range 98%
Specific Gravity 1.36
Linear Formula (CH3)2C6H3Br
Formula Weight 185.06
Percent Purity 98%
Chemical Name or Material 5-Bromo-m-xylene
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-m-xylene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.