missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-DL-tryptophan, 99%, Thermo Scientific Chemicals

Product Code. 10686334
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6548-09-0
Molecular Formula C11H11BrN2O2
Molecular Weight (g/mol) 283.13
MDL Number MFCD00005648
InChI Key KZDNJQUJBMDHJW-UHFFFAOYSA-N
Synonym 5-bromo-dl-tryptophan, 2-amino-3-5-bromo-1h-indol-3-yl propanoic acid, 5-bromotryptophan, tryptophan, 5-bromo, 5-bromo-tryptophan, 2-amino-3-5-bromoindol-3-yl propanoic acid, acmc-20a9ck, 2-amino-3-5-bromo-1h-indol-3-yl-propionic acid, 5-bromo-dl-tryptophan, sr-2-amino-3-5-bromoindolyl propionic acid
PubChem CID 96735
IUPAC Name 2-amino-3-(5-bromo-1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC=C(Br)C=C12)C(O)=O

Specifications

Melting Point 261.0°C to 267.0°C
CAS Min % 98.5
CAS Max % 100.0
Color White to Beige
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 283.12
Percent Purity 99%
Quantity 5 g
Physical Form Fine Crystalline Powder
Chemical Name or Material 5-Bromo-DL-tryptophan
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'