missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-8-methoxyquinoline, 96%

Product Code. 15400698
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400698 250 mg 250mg
15480688 1 g 1g
15490688 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400698 Supplier Thermo Scientific Alfa Aesar Supplier No. H54386.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10522-47-1 | C10H8BrNO | 238.08 g/mol

5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10522-47-1
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.08
MDL Number MFCD04966996
InChI Key ZRQISUREPLHYIG-UHFFFAOYSA-N
Synonym quinoline, 5-bromo-8-methoxy
PubChem CID 4715023
IUPAC Name 5-bromo-8-methoxyquinoline
SMILES COC1=C2N=CC=CC2=C(Br)C=C1

Specifications

Melting Point 80°C to 82°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 238.08
Percent Purity 96%
Chemical Name or Material 5-Bromo-8-methoxyquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.