missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-7-nitroindole, 99%, Thermo Scientific™

Product Code. 11393726
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11393726

Brand: Thermo Scientific Alfa Aesar L14923.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-7-nitroindole is used in gas chromatography analysis. It is also used as pharmaceutical and medical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165669-16-9
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.04
MDL Number MFCD00619260
InChI Key WERRDKWLHFDNTO-UHFFFAOYSA-N
Synonym 5-bromo-7-nitroindole, acmc-20a3m9
PubChem CID 816685
IUPAC Name 5-bromo-7-nitro-1H-indole
SMILES [O-][N+](=O)C1=C2NC=CC2=CC(Br)=C1

Specifications

Melting Point 211°C to 214°C
Quantity 1 g
Beilstein 8139934
Solubility Information Insoluble in water.
Formula Weight 241.05
Percent Purity 99%
Chemical Name or Material 5-Bromo-7-nitroindole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.