missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-6-methylpyridin-2-amine, 97%, Thermo Scientific™

Product Code. 10102994
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 42753-71-9
Molecular Formula C6H8BrN2
Molecular Weight (g/mol) 188.05
MDL Number MFCD00068230
InChI Key SEOZHXRTVJPQPZ-UHFFFAOYSA-O
Synonym 2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine
PubChem CID 170678
IUPAC Name 5-bromo-6-methylpyridin-2-amine
SMILES CC1=[NH+]C(N)=CC=C1Br

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 187.04
Percent Purity 95%
Chemical Name or Material 5-Bromo-6-methylpyridin-2-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'