missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-methyl-2-phenyl-1,3-oxazole, 95%, Thermo Scientific™

Product Code. 10152974
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10152974 5 g Amber Glass Bottle 5g
10526294 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10152974 Supplier Thermo Scientific Maybridge Supplier No. CC18110DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 21354-98-3
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.08
MDL Number MFCD11109319
InChI Key QHQQHNFHCQSTBJ-UHFFFAOYSA-N
Synonym 5-bromo-4-methyl-2-phenyloxazole, 4-methyl-2-phenyl-5-bromooxazole, oxazole,5-bromo-4-methyl-2-phenyl, 5-bromo-4-methyl-1,3-oxazol-2-yl benzene
PubChem CID 15328586
IUPAC Name 5-bromo-4-methyl-2-phenyl-1,3-oxazole
SMILES CC1=C(Br)OC(=N1)C1=CC=CC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 238.08
Percent Purity 95%
Chemical Name or Material 5-Bromo-4-methyl-2-phenyl-1, 3-oxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.