missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole, ≥97%, Thermo Scientific™

Product Code. 10320913
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10320913 10 g Amber Glass Bottle 10g
10709923 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10320913 Supplier Thermo Scientific Maybridge Supplier No. MO00859EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 850375-27-8
Molecular Formula C11H7BrF3NS
Molecular Weight (g/mol) 322.143
MDL Number MFCD04974053
InChI Key BZCIYGNYGIKEEC-UHFFFAOYSA-N
Synonym 5-bromo-4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole, 5-bromo-4-methyl-2-4-trifluoromethyl-phenyl-1,3-thiazole, 5-bromo-4-methyl-2-4-trifluoromethyl phenyl thiazole, 5-bromanyl-4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole
PubChem CID 2824063
IUPAC Name 5-bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 322.15
Percent Purity 97%
Chemical Name or Material 5-Bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.