missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl Phosph., p-Toluidine Salt (Wht. to Off-Wht. Powd.), Fisher BioReagents

Product Code. 10619553
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Amber Glass
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10619553 500 mg Amber Glass 500mg
10183053 100 mg Amber Glass 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10619553 Supplier Fisher BioReagents Supplier No. BP1610500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
BCIP is used in conjunction with Nitro Blue Tetrazolium (NBT) in the colorimetric detection of Alkaline Phosphatase. It is suitable for use in immunohistochemistry, immunoblot staining, and ELISA applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H15BrClN2O4P
Molecular Weight (g/mol) 433.62
InChI Key QEIFSLUFHRCVQL-UHFFFAOYSA-N
PubChem CID 81062
IUPAC Name 4-nitro-2,1,3-benzothiadiazole
SMILES CC1=CC=C(N)C=C1.OP(O)(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12

Specifications

Alpha Vector Bromo-4-chloro-3-indolyl Phosphate,p-Toluidine Salt (BCIP)
CAS 1708766
Melting Point 222.8°C
Color White
Assay Percent Range ≥99 %
Packaging Amber Glass
Quantity 500 mg
Synonym 4-nitrobenzo c 1,2,5 thiadiazole, 4-nitropiazthiole, 2,1,3-benzothiadiazole, 4-nitro, 4-nitro-1,2,3-benzothiadiazole, 4-nitro-benzo 1,2,5 thiadiazole, 4-nitro-2??,1,3-benzothiadiazole, 4-nitrobenzo c 1,2,5-thiadiazole, d0v8fg, acmc-1b59l, 4-bromo-2,8-dimethylphenol
Formula Weight 433.62
Percent Purity 99% min.
Physical Form Solid
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl Phosphate, p-Toluidine Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.