missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl Phosph., p-Toluidine Salt (Wht. to Off-Wht. Powd.), Fisher BioReagents

Catalog Number 10183053
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Amber Glass
This item is not returnable. View return policy
C15H15BrClN2O4P
433.62
QEIFSLUFHRCVQL-UHFFFAOYSA-N
This item is not returnable. View return policy
C15H15BrClN2O4P
433.62
QEIFSLUFHRCVQL-UHFFFAOYSA-N
BCIP is used in conjunction with Nitro Blue Tetrazolium (NBT) in the colorimetric detection of Alkaline Phosphatase. It is suitable for use in immunohistochemistry, immunoblot staining, and ELISA applications.
TRUSTED_SUSTAINABILITY
Alpha Vector Bromo-4-chloro-3-indolyl Phosphate,p-Toluidine Salt (BCIP)
CAS 6578-06-9
Melting Point 222.8°C
Color White
Assay Percent Range ≥99 %
Packaging Amber Glass
Quantity 100 mg
Synonym BCIP
Formula Weight 433.62
Percent Purity 99% min.
Physical Form Solid
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl Phosphate, p-Toluidine Salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'