missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-chloro-3-indolyl Beta-D-glucuronide, Cyclohexylammonium Salt, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 5-Bromo-4-chloro-3-indolyl beta-D-glucuronide, Cyclohexylammonium salt |
| CAS | 114162-64-0 |
| Molecular Formula | C14 H13 Br Cl N O7 . C6 H13 N |
| Quantity | 100 mg |
| Synonym | β-D-Glucopyranosiduronic acid, 5-bromo-4-chloro-1H-indol-3-yl, compd. with cyclohexanamine (1:1), X-Gluc, 5-Bromo-4-chloro-3-indolyl β-D-glucuronide Cyclohexylammonium Salt |
| InChI Formula | InChI=1S/C14H13BrClNO7.C6H13N/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;7-6-4-2-1-3-5-6/h1-3,9-12,14,17-20H,(H,21,22);6H,1-5,7H2/t9-,10-,11+,12-,14+;/m0./s1 |
| SMILES | NC1CCCCC1.O[C@@H]2[C@@H](O)[C@H](Oc3c[nH]c4ccc(Br)c(Cl)c34)O[C@@H]([C@H]2O)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine |
| Molecular Weight (g/mol) | 521.79 |
| Formula Weight | 520.0612 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction