missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt hydrate, 98%

Product Code. 10072001 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072001 100 mg Glass bottle 100mg
10628364 25 mg Glass bottle 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10072001 Supplier Thermo Scientific Acros Supplier No. 331191000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114162-64-0 | C20H26BrClN2O7 | 521.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114162-64-0
Molecular Formula C20H26BrClN2O7
Molecular Weight (g/mol) 521.79
MDL Number MFCD13185253
InChI Key JXCKZXHCJOVIAV-VLQIOUPANA-N
Synonym 5-bromo-4-chloro-3-indolyl, a-d-glucuronide cyclohe, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
PubChem CID 126842357
IUPAC Name (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium;hydrate
SMILES NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230°C
Color Colorless or White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: soluble
Formula Weight 521.79
Percent Purity 98%
Physical Form Crystals, Powder or Needles
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt hydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.