missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog Number 10628364
Click to view available options
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
114162-64-0
C20H26BrClN2O7
521.79
MFCD13185253
JXCKZXHCJOVIAV-VLQIOUPANA-N
5-bromo-4-chloro-3-indolyl, a-d-glucuronide cyclohe, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
126842357
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium;hydrate
NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O
This item is not returnable. View return policy
114162-64-0
C20H26BrClN2O7
521.79
MFCD13185253
JXCKZXHCJOVIAV-VLQIOUPANA-N
5-bromo-4-chloro-3-indolyl, a-d-glucuronide cyclohe, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
126842357
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium;hydrate
NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 230°C
Color Colorless or White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 mg
Solubility Information Solubility in water: soluble
Formula Weight 521.79
Percent Purity 98%
Physical Form Crystals, Powder or Needles
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt hydrate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'